Learn More
CCDC56 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 204271AP
Catalog No. 50-173-2884
Add to cart
Specifications
Specifications
CCDC56
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9D2R6, Q9Y2R0
CCDC56, HSPC009
Rabbit
IgG
150 μL
-20°C
Primary
28958, 498000, 52469
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
COA3
Liquid
COA3
CCDC56 Fusion Protein Ag14260
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat