Learn More
Abnova™ CCDC6 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00008030T02
Catalog No. 89-938-296
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CCDC6 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 53.3kDa
Transient overexpression cell lysate was tested with anti-CCDC6 antibody (Mfr. No. H00008030-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
BC064391.1
pCMV-CCDC6 full-length
100 μL
Human