Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553904
Description
Immunogen sequence: QPVSAPPSPR DISMEIDSPE NMMRHIRFLK NEVERLKKQL RAAQLQHSEK MAQYLEEERH MREENLRLQR KLQREMERRE ALCRQLSESE SSLEMDDERY FNEMSAQGLR PR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a coiled-coil domain-containing protein. The encoded protein is ubiquitously expressed and may function as a tumor suppressor. A chromosomal rearrangement resulting in the expression of a fusion gene containing a portion of this gene and the intracellular kinase-encoding domain of the ret proto-oncogene is the cause of thyroid papillary carcinoma.
Specifications
CCDC6 | |
Polyclonal | |
Unconjugated | |
CCDC6 | |
2810012H18Rik; AA536681; AA960498; AW061011; CCDC6; coiled-coil domain containing 6; coiled-coil domain-containing protein 6; D10S170; H4; Papillary thyroid carcinoma-encoded protein; Protein H4; PTC; TPC; TST1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8030 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q16204 | |
CCDC6 | |
Recombinant protein corresponding to Human CCDC6. Recombinant protein control fragment (Product #RP-92556). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction