Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC63 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548222

Catalog No. PA548222

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Plays a role in spermiogenesis. Involved in the elongation of flagella and the formation of sperm heads.
Specifications

Specifications

CCDC63
Polyclonal
PBS with 0.09% sodium azide
Q8NA47
Coiled-coil domain-containing protein 63, CCDC63
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
160762
Flow Cytometry, Western Blot
Unconjugated
CCDC63
Liquid
CCDC63
KLH conjugated synthetic peptide between 532-560 amino acids from the C-terminal region of human CCDC63.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit