Learn More
Aviva Systems Biology Corp CCDC70 Antibody - middle region (ARP53804_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53804_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCDC70. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCDC70. Alternate names: DKFZP434K1172, FLJ25853ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-61133