Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC72 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 203931AP150UL
Description
CCDC72, also named as TMA7, is a novel modulator for hair follicle development. CCDC72 has the promoter activity and regulation of gene expression.Specifications
CCDC72 | |
Polyclonal | |
Unconjugated | |
TMA7 | |
CCDC72, HSPC016, HSPC330, TMA7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51372, 66167, 679747 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K003, Q9Y2S6 | |
TMA7 | |
CCDC72 Fusion Protein Ag13792 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction