Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPATCH11 Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548967

Catalog No. PA548967

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

GPATCH11
Polyclonal
PBS with 0.09% sodium azide
Q8N954, Q3UFS4
G patch domain-containing protein 11, Coiled-coil domain-containing protein 75, GPATCH11, CCDC75
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100765159, 253635, 53951
Western Blot
Unconjugated
GPATCH11
Liquid
GPATCH11
KLH conjugated synthetic peptide between 12-41 amino acids from the N-terminal region of human CCDC75.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit