Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC85A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560241
Description
Immunogen sequence: RHPHPGSSPE TLPKHVLSGS PEHFQKHRSG SSPEHARHSG GSPEHLQKHA LGGSLEHLPR ARGTSPEHLK QHYGGSPDHK HGGGSGGS Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
CCDC85A is a protein coding gene.
Specifications
CCDC85A | |
Polyclonal | |
Unconjugated | |
CCDC85A | |
Ccdc85a; coiled-coil domain containing 85A; coiled-coil domain-containing protein 85A; KIAA1912; RGD1311700; zgc:175167 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114800 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PX6 | |
CCDC85A | |
Recombinant protein corresponding to Human CCDC85A. Recombinant protein control fragment (Product #RP-99636). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction