Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ccdc89 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549322

Catalog No. PA549322

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Ccdc89
Polyclonal
PBS with 0.09% sodium azide
Q8N998, Q9DA73, B2RZ86
Coiled-coil domain-containing protein 89, Bc8 orange-interacting protein, Ccdc89 {ECO:0000312|EMBL:AAI670631, ECO:0000312|RGD:1305335}
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
220388, 293107, 70054
Western Blot
Unconjugated
CCDC89
Liquid
CCDC89
KLH conjugated synthetic peptide between 114-145 amino acids from the Central region of rat Ccdc89.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit