Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC91 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578525
Description
Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Pig (94%), Bovine (93%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CCDC91 is involved in the regulation of membrane traffic through the trans-Golgi network (TGN).
Specifications
CCDC91 | |
Polyclonal | |
Unconjugated | |
CCDC91 | |
1700086G08Rik; 1810060J02Rik; AU045683; AW555238; Ccdc91; coiled-coil domain containing 91; coiled-coil domain-containing protein 91; GGA binding partner; GGA-binding partner; Ggabp; HSD8; p56; p56 accessory protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55297 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M Tris glycine with 20% glycerol, 0.1M glycine and 0.01% thimerosal; pH 7 | |
Q7Z6B0 | |
CCDC91 | |
Recombinant protein encompassing a sequence within the center region of human CCDC91. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction