Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC91 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548924

Catalog No. PA548924

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in the regulation of membrane traffic through the trans-Golgi network (TGN).
Specifications

Specifications

CCDC91
Polyclonal
PBS with 0.09% sodium azide
Q7Z6B0, Q9D8L5
Coiled-coil domain-containing protein 91, GGA-binding partner, p56 accessory protein, CCDC91, GGABP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55297, 67015
Western Blot
Unconjugated
CCDC91
Liquid
CCDC91
KLH conjugated synthetic peptide between 322-350 amino acids from the C-terminal region of human CCDC91.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit