Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC94 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen CCDC94
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126356
View Documents
Novus Biologicals
NBP31072725UL
25 μg
Each for $106.19
Add to cart
 
NB126355
View Documents
Novus Biologicals
NBP310727100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

CCDC94 Polyclonal antibody specifically detects CCDC94 in Human samples. It is validated for Western Blot
Specifications

Specifications

CCDC94
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
55702
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Human
coiled-coil domain containing 94, coiled-coil domain-containing protein 94, FLJ10374
The immunogen is a synthetic peptide directed towards the C-terminal region of Human CCDC94 (NP_060544). Peptide sequence KPKRKVEVWEQSVGSLGSRPPLSRLVVVKKAKADPDCSNGQPQAAPTPGA
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit