Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPDL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548586

Catalog No. PA548586

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with monkey based on sequence homology.

Required for the localization of dynein and dynactin to the mitotic kintochore. Dynein is believed to control the initial lateral interaction between the kinetochore and spindle microtubules and to facilitate the subsequent formation of end-on kinetochore-microtubule attachments mediated by the NDC80 complex. Also required for correct spindle orientation. Does not appear to be required for the removal of spindle assembly checkpoint (SAC) proteins from the kinetochore upon bipolar spindle attachment.
Specifications

Specifications

SPDL1
Polyclonal
PBS with 0.09% sodium azide
Q96EA4
Protein Spindly, hSpindly, Arsenite-related gene 1 protein, Coiled-coil domain-containing protein 99, Rhabdomyosarcoma antigen MU-RMS-404A, Spindle apparatus coiled-coil domain-containing protein 1, SPDL1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54908
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SPDL1
Liquid
SPDL1
KLH conjugated synthetic peptide between 459-488 amino acids from the C-terminal region of human CCDC99.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit