Tocris™ CCG 203971

Inhibits the MRTF/SRF gene transcription pathway; antifibrotic agent

Manufacturer: Tocris™ 5277/10

 View more versions of this product

Catalog No. 52-771-0

  N/A
Add to Cart

Disclaimers:

For Research Use Only

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Inhibitors CCG 203971
    Purity >98
    Molecular Weight 408.88
    Quantity 10mg
    Product Type CCG 203971

    Antifibrotic agent; inhibits fibrosis by targeting the MRTF/SRF gene transcription pathway. Selectively inhibits proliferation of SSc-derived dermal fibroblasts but not that of normal fibroblasts. Inhibits expression of CTGF, α-SMA, and COL1A2 in SSc fibroblasts as well as in LPA and in TGFβ-stimulated fibroblasts. Prevents bleomycin-induced skin thickening and collagen deposition in vivo. Also suppresses PC-3 cell migration in scratch wound assays (IC50 = 4.2µM).

    • Chemical Name: N-(4-Chlorophenyl)-1-[3-(2-furanyl)benzoyl]-3-piperidinecarboxamide
    • Formula: C23H21ClN2O3
    • CAS Number: 1443437-74-8
    • Solubility: Soluble to 50mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit