Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCG 2046, Tocris Bioscience™

Catalog Number p-4783834
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
13017-69-1
C11H10N4
198.229
OUAQSPOCQDQFEV-UHFFFAOYSA-N
1,1,2,2-tetracyano-3-methyl-3-propylcyclopropane, 1,1,2,2-cyclopropanetetracarbonitrile,methylpropyl, 3-methyl-3-propyl-1,1,2,2-cyclopropanetetracarbonitrile
263681
3-methyl-3-propylcyclopropane-1,1,2,2-tetracarbonitrile
CCCC1(C(C1(C#N)C#N)(C#N)C#N)C
This item is not returnable. View return policy
13017-69-1
C11H10N4
198.229
OUAQSPOCQDQFEV-UHFFFAOYSA-N
1,1,2,2-tetracyano-3-methyl-3-propylcyclopropane, 1,1,2,2-cyclopropanetetracarbonitrile,methylpropyl, 3-methyl-3-propyl-1,1,2,2-cyclopropanetetracarbonitrile
263681
3-methyl-3-propylcyclopropane-1,1,2,2-tetracarbonitrile
CCCC1(C(C1(C#N)C#N)(C#N)C#N)C

Inhibitor of regulator of G-protein signaling 4 (RGS4)

Inhibitor of regulator of G-protein signaling 4 (RGS4). Reduces the RGS4-Gαo protein-protein interaction signal.
Quantity 10 mg
Formula Weight 198.22
Percent Purity >99%
Chemical Name or Material CCG 2046
Recommended Storage Desiccate at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit