Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCG 50014, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
883050-24-6
C16H13FN2O2S
316.35
QUIIIYITNGOFEI-UHFFFAOYSA-N
compound 1a,unii-ra72g28ve9,4-4-fluorobenzyl-2-p-tolyl-1,2,4-thiadiazolidine-3,5-dione,4-4-fluorophenyl methyl-2-4-methylphenyl-1,2,4-thiadiazolidine-3,5-dione,cyto9f11,maybridge1_007327,d0c2gr,4-4-fluorobenzyl-2-4-methylphenyl-1,2,4-thiadiazolane-3,5-dione,4-4-fluorophenyl methyl-2-p-tolyl-1,2,4-thiadiazolidine-3,5-dione
2733079
4-[(4-fluorophenyl)methyl]-2-(4-methylphenyl)-1,2,4-thiadiazolidine-3,5-dione
CC1=CC=C(C=C1)N2C(=O)N(C(=O)S2)CC3=CC=C(C=C3)F
This item is not returnable. View return policy
883050-24-6
C16H13FN2O2S
316.35
QUIIIYITNGOFEI-UHFFFAOYSA-N
compound 1a,unii-ra72g28ve9,4-4-fluorobenzyl-2-p-tolyl-1,2,4-thiadiazolidine-3,5-dione,4-4-fluorophenyl methyl-2-4-methylphenyl-1,2,4-thiadiazolidine-3,5-dione,cyto9f11,maybridge1_007327,d0c2gr,4-4-fluorobenzyl-2-4-methylphenyl-1,2,4-thiadiazolane-3,5-dione,4-4-fluorophenyl methyl-2-p-tolyl-1,2,4-thiadiazolidine-3,5-dione
2733079
4-[(4-fluorophenyl)methyl]-2-(4-methylphenyl)-1,2,4-thiadiazolidine-3,5-dione
CC1=CC=C(C=C1)N2C(=O)N(C(=O)S2)CC3=CC=C(C=C3)F

Potent and selective inhibitor of regulator of G-protein signaling 4 (RGS4) protein

Potent and selective inhibitor of regulator of G-protein signaling 4 (RGS4) protein (IC50 values are 30.1 nM and 11.0 μM for RGS4 and RGS8 respectively). Exhibits >20-fold selectivity for RGS4 over other RGS proteins.
Quantity 50mg
Formula Weight 316.35
Percent Purity >98%
Chemical Name or Material CCG 50014

Recommended Storage : Store at +4°C