Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCG 50014, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective inhibitor of regulator of G-protein signaling 4 (RGS4) protein (IC50 values are 30.1 nM and 11.0 μM for RGS4 and RGS8 respectively). Exhibits >20-fold selectivity for RGS4 over other RGS proteins.
Specifications
Quantity | 10 mg |
Formula Weight | 316.35 |
Percent Purity | >98% |
Chemical Name or Material | CCG 50014 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction