Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCG 63802, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
620112-78-9
C26H18N4O2S
450.516
VFSVKVQMZDJFQX-NBVRZTHBSA-N
unii-339bu4y35u, 2e-2-1,3-benzothiazol-2-yl-3-9-methyl-2-3-methylphenoxy-4-oxo-4h-pyrido 1,2-a pyrimidin-3-yl prop-2-enenitrile, e-2-1,3-benzothiazol-2-yl-3-9-methyl-2-3-methylphenoxy-4-oxopyrido 1,2-a pyrimidin-3-yl prop-2-enenitrile, e-2-benzo d thiazol-2-yl-3-9-methyl-4-oxo-2-m-tolyloxy-4h-pyrido 1,2-a pyrimidin-3-yl acrylonitrile, 2e-2-1,3-benzothiazol-2-yl-3-9-methyl-2-3-methylphenoxy-4-oxopyrido 1,2-a pyrimidin-3-yl prop-2-enenitrile, 2e-2-1,3-benzothiazol-2-yl-3-9-methyl-2-3-methylphenoxy-4-oxo-4h-pyrido 1,2-a pyrimidin-3-yl acrylonitrile, e-2-benzo d thiazol-2-yl-3-9-methyl-4-oxo-2-m-tolyloxy-4h-pyrido 1,2-a pyrimidin-3-yl acrylon
6057678
(E)-2-(1,3-benzothiazol-2-yl)-3-[9-methyl-2-(3-methylphenoxy)-4-oxopyrido[1,2-a]pyrimidin-3-yl]prop-2-enenitrile
CC1=CC(=CC=C1)OC2=C(C(=O)N3C=CC=C(C3=N2)C)C=C(C#N)C4=NC5=CC=CC=C5S4
This item is not returnable. View return policy
620112-78-9
C26H18N4O2S
450.516
VFSVKVQMZDJFQX-NBVRZTHBSA-N
unii-339bu4y35u, 2e-2-1,3-benzothiazol-2-yl-3-9-methyl-2-3-methylphenoxy-4-oxo-4h-pyrido 1,2-a pyrimidin-3-yl prop-2-enenitrile, e-2-1,3-benzothiazol-2-yl-3-9-methyl-2-3-methylphenoxy-4-oxopyrido 1,2-a pyrimidin-3-yl prop-2-enenitrile, e-2-benzo d thiazol-2-yl-3-9-methyl-4-oxo-2-m-tolyloxy-4h-pyrido 1,2-a pyrimidin-3-yl acrylonitrile, 2e-2-1,3-benzothiazol-2-yl-3-9-methyl-2-3-methylphenoxy-4-oxopyrido 1,2-a pyrimidin-3-yl prop-2-enenitrile, 2e-2-1,3-benzothiazol-2-yl-3-9-methyl-2-3-methylphenoxy-4-oxo-4h-pyrido 1,2-a pyrimidin-3-yl acrylonitrile, e-2-benzo d thiazol-2-yl-3-9-methyl-4-oxo-2-m-tolyloxy-4h-pyrido 1,2-a pyrimidin-3-yl acrylon
6057678
(E)-2-(1,3-benzothiazol-2-yl)-3-[9-methyl-2-(3-methylphenoxy)-4-oxopyrido[1,2-a]pyrimidin-3-yl]prop-2-enenitrile
CC1=CC(=CC=C1)OC2=C(C(=O)N3C=CC=C(C3=N2)C)C=C(C#N)C4=NC5=CC=CC=C5S4

Inhibitor of regulator of G-protein signaling 4 (RGS4) protein

Reversible inhibitor of regulator of G-protein signaling (RGS) proteins. Selective amongst RGS proteins, with greatest potency at RGS4. Inhibits GTPase accelerating protein activity of RGS4 and blocks its interaction with Gαo. Retains activity under reducing conditions.
Quantity 10 mg
Formula Weight 450.51
Percent Purity >99%
Chemical Name or Material CCG 63802

Recommended Storage : Store at +4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit