Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL12/MCP-5 Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0169-8J25, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212089AF532
Description
CCL12/MCP-5 Monoclonal specifically detects CCL12/MCP-5 in Mouse samples. It is validated for Western Blot.Specifications
CCL12/MCP-5 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Ccl12 | |
Mouse recombinant CCL12/MCP-5 | |
0.1 mL | |
20293 | |
Store at 4°C in the dark. |
Western Blot | |
RM0169-8J25 | |
Western Blot | |
C-C motif chemokine 12, chemokine (C-C motif) ligand 12, MCP-5, Scya12 | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title