Learn More
CCL13/MCP-4 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0137-3G14, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212090AF594
Catalog No. NP212090A94
Add to cart
Description
Description
CCL13/MCP-4 Monoclonal antibody specifically detects CCL13/MCP-4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CCL13/MCP-4
MM0137-3G14
Flow Cytometry
C-C motif chemokine 13, chemokine (C-C motif) ligand 13, CKb10, MCP4, MCP-4Monocyte chemoattractant protein 4, MGC17134, NCC-1Monocyte chemotactic protein 4, NCC1Small-inducible cytokine A13, new CC chemokine 1, SCYA13CK-beta-10, SCYL1, small inducible cytokine subfamily A (Cys-Cys), member 13
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CCL13
Human MCP4
0.1 mL
Primary
6357