Learn More
CCL15/MIP-1 delta Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0139-2M24, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212092AF750
Catalog No. NP212092A70
Add to cart
Description
Description
CCL15/MIP-1 delta Monoclonal antibody specifically detects CCL15/MIP-1 delta in Human samples. It is validated for Western Blot.
Specifications
Specifications
CCL15/MIP-1 delta
MM0139-2M24
Western Blot
chemokine (C-C motif) ligand 15, Chemokine CC-2, HCC-2Lkn-1, leukotactin 1, Leukotactin-1, LKN1, LKN-1, Macrophage inflammatory protein 5, MIP-1 delta, MIP5, MIP-5MIP-1d, Mrp-2b, NCC-3MIP-1D, NCC3MRP-2B, new CC chemokine 3, SCYA15member 15, Small-inducible cytokine A15
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CCL15
Human Macrophage Inflammatory Protein 5
0.1 mL
Primary
6359