Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CCL16/HCC-4 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF802SP
This item is not returnable.
View return policy
Description
CCL16/HCC-4/LEC Polyclonal specifically detects CCL16/HCC-4/LEC in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
CCL16/HCC-4/LEC | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-C motif chemokine 16, chemokine (C-C motif) ligand 16, Chemokine CC-4, Chemokine LEC, CKb12, HCC4, HCC-4, HCC-4Liver-expressed chemokine, LCC-1Lymphocyte and monocyte chemoattractant, LEC, liver CC chemokine-1, LMC, LMCSmall-inducible cytokine A16, monotactin-1, MTN-1, NCC-4, NCC4IL-10-inducible chemokine, NCC-4ILINCK, new CC chemokine 4, SCYA16MGC117051, SCYL4, small inducible cytokine subfamily A (Cys-Cys), member 16 | |
Goat | |
Affinity Purified | |
RUO | |
6360 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 4-16 ug/mL | |
O15467 | |
CCL16 | |
E. coli-derived recombinant human CCL16/HCC-4 Gln24-Gln120 Accession # O15467 | |
25 μg | |
Primary | |
Detects human CCL16/HCC-4 in direct ELISAs and Western blots. | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction