Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL16/HCC-4/LEC Mouse anti-Human, Alexa Fluor 488, Clone: 70218, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems IC328G100UG
This item is not returnable.
View return policy
Description
CCL16/HCC-4/LEC Monoclonal specifically detects CCL16/HCC-4/LEC in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
CCL16/HCC-4/LEC | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
O15467 | |
CCL16 | |
E. coli-derived recombinant human CCL16/HCC-4 Gln24-Gln120 Accession # O15467 | |
RUO | |
6360 | |
Human | |
IgG1 |
Flow Cytometry | |
70218 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
C-C motif chemokine 16, chemokine (C-C motif) ligand 16, Chemokine CC-4, Chemokine LEC, CKb12, HCC4, HCC-4, HCC-4Liver-expressed chemokine, LCC-1Lymphocyte and monocyte chemoattractant, LEC, liver CC chemokine-1, LMC, LMCSmall-inducible cytokine A16, monotactin-1, MTN-1, NCC-4, NCC4IL-10-inducible chemokine, NCC-4ILINCK, new CC chemokine 4, SCYA16MGC117051, SCYL4, small inducible cytokine subfamily A (Cys-Cys), member 16 | |
Mouse | |
100 μg | |
Primary | |
Detects human CCL16/HCC-4 in direct ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction