Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCL16 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547948
Description
Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
CCL16 gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. CCL16 displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. This cytokine also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. The expression of this gene is upregulated by IL-10.
Specifications
CCL16 | |
Polyclonal | |
Unconjugated | |
CCL16 | |
C-C motif chemokine 16; C-C motif chemokine ligand 16; CCL16; chemokine (C-C motif) ligand 16; Chemokine CC-4; Chemokine LEC; CKb12; HCC-4; IL-10-inducible chemokine; ILINCK; LCC-1; LEC; liver CC chemokine-1; liver-expressed chemokine; LMC; Lymphocyte and monocyte chemoattractant; MGC117051; monotactin-1; MTN-1; NCC4; NCC-4; new CC chemokine 4; SCYA16; SCYL4; small inducible cytokine subfamily A (Cys-Cys), member 16; small-inducible cytokine A16 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6360 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry, Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O15467 | |
CCL16 | |
E. coli-derived recombinant human CCL16/HCC-4 Gln24-Gln120. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction