Learn More
CCL17 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 223421AP
Catalog No. 50-173-3840
Add to cart
Description
Description
TARC ligand (CCL17) belongs to the CC chemokine family. It is a small cytokine and the gene is located on q arm of chromosome 16, in humans, along with other chemokines. TARC specifically binds to T-cells and interacts with chemokine receptors CCR4 and CCR8. It plays an important role in T cell development in thymus as well as in trafficking and activation of mature T cells.
Specifications
Specifications
CCL17
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q92583
A 152E5.3, ABCD 2, C C motif chemokine 17, CC chemokine TARC, CCL17, SCYA17, Small inducible cytokine A17, TARC
Rabbit
IgG
150 μL
-20°C
Primary
6361
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CCL17
Liquid
CCL17
CCL17 Fusion Protein Ag17832
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human