Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL17/TARC Antibody (54015), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3641K
Description
CCL17/TARC Monoclonal antibody specifically detects CCL17/TARC in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
CCL17/TARC | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
54015 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARCA-152E5.3, Thymus and activation-regulated chemokine | |
E. coli-derived recombinant human CCL17/TARC, Ala24-Ser94, Accession # Q92583.1 | |
0.5 mL | |
Biologically Active Proteins, Chemokines and Cytokines | |
6361 | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction