Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ CCL17/TARC Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
$832.00
Specifications
Assay Range | 15.625 pg/mL |
---|---|
Assay Sensitivity | 15.625-1000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Immunoassay Kit Format | Sandwich ELISA Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB010121
|
Novus Biologicals™
NBP27954115PLATES |
15 Plates |
Each of 1 for $832.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CCL17/TARC.Specifications
15.625 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CCL17/TARC. | |
Sandwich ELISA | |
6361 | |
Mouse | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1000 pg/mL is recommended. |
15.625-1000 pg/mL | |
HRP | |
Antibody Pairs | |
Human | |
ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARCA-152E5.3, Thymus and activation-regulated chemokine | |
CCL17 | |
Biologically Active Proteins, Chemokines and Cytokines | |
CCL17/TARC |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title