Learn More
CCL17/TARC Rat anti-Mouse, Alexa Fluor™ 594, Clone: RM0170-9K49, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212095AF594
Catalog No. NP21209A94
Add to cart
Description
Description
CCL17/TARC Monoclonal antibody specifically detects CCL17/TARC in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
CCL17/TARC
RM0170-9K49
Western Blot
ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARCA-152E5.3, Thymus and activation-regulated chemokine
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CCL17
Mouse TARC
0.1 mL
Primary
6361