Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL19/MIP-3 beta Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI2A12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271278AF405

Catalog No. N271278A405


Add to cart

Description

Description

CCL19/MIP-3 beta Monoclonal antibody specifically detects CCL19/MIP-3 beta in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

CCL19/MIP-3 beta
OTI2A12
Immunohistochemistry
Affinity Purified, Conjugated
CCL19
Recombinant protein fragment corresponding to amino acids 22-98 of human CCL19/MIP-3 beta (NP_006265) produced in E.coli.
Immunogen affinity purified
Biologically Active Proteins
Primary
6363.0
Immunohistochemistry
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
beta chemokine exodus-3, Beta-chemokine exodus-3, CC chemokine ligand 19, C-C motif chemokine 19, chemokine (C-C motif) ligand 19, CKb11, EBI1-ligand chemokine, ELCMIP-3-beta, Epstein-Barr virus-induced molecule 1 ligand chemokine, exodus-3, Macrophage inflammatory protein 3 beta, macrophage inflammatory protein 3-beta, MGC34433, MIP-3b, MIP3BCK beta-11, SCYA19EBI1 ligand chemokine, small inducible cytokine subfamily A (Cys-Cys), member 19, Small-inducible cytokine A19
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit