Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL19/MIP-3 beta Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0143-11B4, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212097AF594
Description
CCL19/MIP-3 beta Monoclonal specifically detects CCL19/MIP-3 beta in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CCL19/MIP-3 beta | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CCL19 | |
Human recombinant CCL19/MIP-3 beta | |
Primary | |
Human | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
MM0143-11B4 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
beta chemokine exodus-3, Beta-chemokine exodus-3, CC chemokine ligand 19, C-C motif chemokine 19, chemokine (C-C motif) ligand 19, CKb11, EBI1-ligand chemokine, ELCMIP-3-beta, Epstein-Barr virus-induced molecule 1 ligand chemokine, exodus-3, Macrophage inflammatory protein 3 beta, macrophage inflammatory protein 3-beta, MGC34433, MIP-3b, MIP3BCK beta-11, SCYA19EBI1 ligand chemokine, small inducible cytokine subfamily A (Cys-Cys), member 19, Small-inducible cytokine A19 | |
Mouse | |
0.1 mL | |
6363 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title