Learn More
CCL2/JE/MCP-1 Rabbit anti-Human, Alexa Fluor 532, Clone: 144, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP289290AF532
Catalog No. NB066118
Add to cart
Description
Description
CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications
Specifications
CCL2/JE/MCP-1
144
ELISA, Sandwich ELISA Detection
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor 532
50mM Sodium Borate
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, Mcp-1, SCYA2, SMC-CF
This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL2/JE/MCP-1 (NP_002973.1; Gln24-Thr99).
Protein A purified
RUO
Primary
6347