Learn More
CCL20/MIP-3 alpha Mouse anti-Human, Alexa Fluor™ 594, Clone: 3N2D9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227145AF594
Catalog No. NP22714A94
Add to cart
Description
Description
CCL20/MIP-3 alpha Monoclonal antibody specifically detects CCL20/MIP-3 alpha in Human samples. It is validated for Western Blot.
Specifications
Specifications
CCL20/MIP-3 alpha
3N2D9
Western Blot
beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CCL20
Full length recombinant human MIP-3 was used as immunogen for this antibody.
0.1 mL
Primary
6364