Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL21/6Ckine Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0174-10K10, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212104AF532
Description
CCL21/6Ckine Monoclonal specifically detects CCL21/6Ckine in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCL21/6Ckine | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CCL21 | |
Mouse recombinant CCL21/6Ckine | |
0.1 mL | |
6366 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0174-10K10 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, member 21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLCSecondary lymphoid-tissue chemokine | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title