Learn More
CCL22/MDC Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0147-13G44, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212105AF594
Catalog No. NP21210A94
Add to cart
Description
Description
CCL22/MDC Monoclonal antibody specifically detects CCL22/MDC in Human samples. It is validated for Western Blot.
Specifications
Specifications
CCL22/MDC
MM0147-13G44
Western Blot
A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CCL22
Human MDC
0.1 mL
Primary
6367