Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL22/MDC Mouse anti-Human, Clone: 6F5.7, MilliporeSigma™

Mouse Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen CCL22/MDC
Clone 6F5.7
Concentration 1 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABC1703100
View Documents
MilliporeSigma
MABC1703100UG
100 μg
Each for $373.00
Add to cart
 
MABC170325
View Documents
MilliporeSigma
MABC170325UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

CCL22/MDC Monoclonal antibody specifically detects CCL22/MDC in Human samples. It is validated for Western Blotting, Immunohistochemistry (Paraffin).
Specifications

Specifications

CCL22/MDC
1 mg/mL
Unconjugated
Mouse
Human
Recombinant fragment of human CCL22 corresponding to 67 amino acids in the mature protein.
Monoclonal
Protein G purified
6F5.7
Western Blot, Immunohistochemistry (Paraffin)
Purified
RUO
Purified mouse monoclonal antibody IgG2a in PBS with 0.05% sodium azide.
IgG2aκ
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit