Learn More
CCL22/MDC Rat anti-Mouse, Alexa Fluor 700, Clone: RM0175-9L23, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212106AF700
Catalog No. NB212106700
Add to cart
Description
Description
CCL22/MDC Monoclonal specifically detects CCL22/MDC in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
CCL22/MDC
RM0175-9L23
Western Blot
Purified
CCL22
Mouse MDC
Protein G purified
Biologically Active Proteins
Primary
6367
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse