Learn More
CCL23/Ck beta 8-1/MIP3 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0145-3K21, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212101AF532
Catalog No. NP212101A32
Add to cart
Description
Description
CCL23/Ck beta 8-1/MIP3 Monoclonal antibody specifically detects CCL23/Ck beta 8-1/MIP3 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CCL23/Ck beta 8-1/MIP3
MM0145-3K21
Western Blot
C6 beta-chemokine, chemokine (C-C motif) ligand 23, CKb8, CKB-8, CK-beta-8, hmrp-2a, Macrophage inflammatory protein 3, MIP3, MIP-3CK-BETA-8, MPIF1, MPIF-1Ckb-8-1, Myeloid progenitor inhibitory factor 1, SCYA23C-C motif chemokine 23, small inducible cytokine subfamily A (Cys-Cys), member 23, Small-inducible cytokine A23
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CCL23
Human Macrophage Inflammatory Protein 3
0.1 mL
Primary
6368