Learn More
CCL23/Ck beta 8-1/MIP3 Mouse anti-Human, Alexa Fluor 700, Clone: MM0148-12G51, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212107AF700
Catalog No. NB212107700
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CCL23/Ck beta 8-1/MIP3 Monoclonal antibody specifically detects CCL23/Ck beta 8-1/MIP3 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CCL23/Ck beta 8-1/MIP3
MM0148-12G51
Western Blot
Purified
CCL23
Human Macrophage Inflammatory Protein 3
Protein G purified
Biologically Active Proteins, Cell Cycle and Replication
Primary
6368
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
C6 beta-chemokine, chemokine (C-C motif) ligand 23, Ckb-8, CKb8, CK-beta-8, hmrp-2a, Macrophage inflammatory protein 3, MIP3, MIP-3CK-BETA-8, MPIF1, MPIF-1Ckb-8-1, Myeloid progenitor inhibitory factor 1, SCYA23C-C motif chemokine 23, small inducible cytokine subfamily A (Cys-Cys), member 23, Small-inducible cytokine A23
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human