Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL23/Ck beta 8-1/MIP3 Mouse anti-Human, Clone: MM0148-12G51, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | CCL23/Ck beta 8-1/MIP3 |
---|---|
Clone | MM0148-12G51 |
Concentration | LYOPH |
Dilution | Western Blot 1:100-1:2000, Blocking/Neutralizing 50-200 molar excess |
Applications | Western Blot, Blocking Assay |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP212107
|
Novus Biologicals
NBP212107 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB019050
|
Novus Biologicals
NBP2121070.025MG |
0.025 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
CCL23/Ck beta 8-1/MIP3 Monoclonal antibody specifically detects CCL23/Ck beta 8-1/MIP3 in Human samples. It is validated for Western Blot, Block/Neutralize.Specifications
CCL23/Ck beta 8-1/MIP3 | |
LYOPH | |
Western Blot, Blocking Assay | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication | |
P55773 | |
CCL23 | |
IgG1 | |
Protein G purified |
MM0148-12G51 | |
Western Blot 1:100-1:2000, Blocking/Neutralizing 50-200 molar excess | |
Monoclonal | |
Purified | |
RUO | |
Human | |
6368 | |
Human Macrophage Inflammatory Protein 3 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title