Learn More
CCL23 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 178651AP
Catalog No. 50-173-1839
Add to cart
Description
Description
This gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. This chemokine, a member of the CC subfamily, displays chemotactic activity on resting T lymphocytes and monocytes, lower activity on neutrophils and no activity on activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. In addition, the product of this gene is a potent agonist of the chemokine (C-C motif) receptor 1. Alternative splicing results in multiple transcript variants that encode different isoforms.
Specifications
Specifications
CCL23
0.22 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P55773
C C motif chemokine 23, CCL23, CK BETA 8, Ckb 8, Ckb 8 1, CKb8, MIP 3, MIP3, MPIF 1, MPIF1, SCYA23, Small inducible cytokine A23
Rabbit
IgG
150 μL
-20°C
Primary
6368
Western Blot
Unconjugated
CCL23
Liquid
CCL23
CCL23 Fusion Protein Ag12346
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human