Learn More
CCL24/Eotaxin-2/MPIF-2 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0149-8C72, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212108AF594
Catalog No. NP212108A94
Add to cart
Description
Description
CCL24/Eotaxin-2/MPIF-2 Monoclonal antibody specifically detects CCL24/Eotaxin-2/MPIF-2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CCL24/Eotaxin-2/MPIF-2
MM0149-8C72
Western Blot
chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CCL24
Human Eotaxin 2
0.1 mL
Primary
6369