Learn More
CCL24/Eotaxin-2/MPIF-2 Rat anti-Human, Mouse, Alexa Fluor™ 350, Clone: RM0176-8M17, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212109AF350
Catalog No. NP212109A30
Add to cart
Description
Description
CCL24/Eotaxin-2/MPIF-2 Monoclonal antibody specifically detects CCL24/Eotaxin-2/MPIF-2 in Human,Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
CCL24/Eotaxin-2/MPIF-2
RM0176-8M17
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CCL24
Mouse Eotaxin 2
0.1 mL
Primary
6369