Learn More
CCL25/TECK Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0150-9G99, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212110AF350
Catalog No. NP212110A30
Add to cart
Description
Description
CCL25/TECK Monoclonal antibody specifically detects CCL25/TECK in Human samples. It is validated for Western Blot.
Specifications
Specifications
CCL25/TECK
MM0150-9G99
Western Blot
C-C motif chemokine 25, chemokine (C-C motif) ligand 25, Chemokine TECK, Ck beta-15, Ckb15, SCYA25TECKvar, small inducible cytokine subfamily A (Cys-Cys), member 25, Small-inducible cytokine A25, TECKMGC150327, thymus expressed chemokine, Thymus-expressed chemokine
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CCL25
Human TECK
0.1 mL
Primary
6370