Learn More
CCL25/TECK Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0177-7N28, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212111AF750
Catalog No. NP212111A70
Add to cart
Description
Description
CCL25/TECK Monoclonal antibody specifically detects CCL25/TECK in Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
CCL25/TECK
RM0177-7N28
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
C-C motif chemokine 25, chemokine (C-C motif) ligand 25, Chemokine TECK, Ck beta-15, Ckb15, SCYA25TECKvar, small inducible cytokine subfamily A (Cys-Cys), member 25, Small-inducible cytokine A25, TECKMGC150327, thymus expressed chemokine, Thymus-expressed chemokine
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CCL25
Mouse TECK
0.1 mL
Primary
6370