Learn More
CCL26/Eotaxin-3 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0151-8D34, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212112AF532
Catalog No. NP212112A32
Add to cart
Description
Description
CCL26/Eotaxin-3 Monoclonal antibody specifically detects CCL26/Eotaxin-3 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CCL26/Eotaxin-3
MM0151-8D34
Western Blot
CC chemokine IMAC, C-C motif chemokine 26, chemokine (C-C motif) ligand 26, chemokine N1, eotaxin-3, IMAC, Macrophage inflammatory protein 4-alpha, MGC126714, MIP-4a, MIP-4alpha, SCYA26MIP-4-alpha, small inducible cytokine A26, small inducible cytokine subfamily A (Cys-Cys), member 26, Small-inducible cytokine A26, TSC-1Thymic stroma chemokine-1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CCL26
Human Eotaxin-3
0.1 mL
Primary
10344