Learn More
CCL27/CTACK Rat anti-Mouse, Alexa Fluor 647, Clone: RM0178-6M35, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212113AF647
Catalog No. NB212113647
Add to cart
Description
Description
CCL27/CTACK Monoclonal specifically detects CCL27/CTACK in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CCL27/CTACK
RM0178-6M35
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
CCL27
Mouse CCL27
Protein G purified
Biologically Active Proteins
Primary
10850
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
ALP, C-C motif chemokine 27, chemokine (C-C motif) ligand 27, CTACKSmall-inducible cytokine A27, CTAK, cutaneous T-cell attracting chemokine, Cutaneous T-cell-attracting chemokine, ESKINE, IL-11 Ralpha-locus chemokine, ILCCC chemokine ILC, PESKY, SCYA27IL-11 R-alpha-locus chemokine, skinkine, small inducible cytokine subfamily A (Cys-Cys), member 27
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse