Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCL28 Monoclonal Antibody (15)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530365
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CCL28/SCYA28.
MEC is a secreted CC chemokine expressed primarily by epithelial cells of the bronchioles, salivary gland, mammary gland and colon. MEC signals through the CCR10 receptor, and chemoattracts resting CD4, CD8 T-cells and eosinophils. MEC contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines.
Specifications
CCL28 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9NRJ3 | |
CCL28 | |
Recombinant Human CCL28/SCYA28 Protein (Ile23-Tyr127). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
15 | |
Unconjugated | |
CCL28 | |
CC chemokine CCL28; C-C motif chemokine 28; C-C motif chemokine ligand 28; CCK 1; CCK1; CCK1 protein; CCL 28; Ccl28; chemokine (C-C motif) ligand 28; chemokine (C-C motif) ligand 28 splice variant chi; MEC; Mucosae-associated epithelial chemokine; Protein CCK1; SCYA28; small inducible cytokine A28; small inducible cytokine subfamily A (Cys-Cys), member 28; small-inducible cytokine A28 | |
Mouse | |
Protein A | |
RUO | |
56477 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction