Learn More
CCL28 Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0153-9G34, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212115AF350
Catalog No. NP21211A30
Add to cart
Description
Description
CCL28 Monoclonal antibody specifically detects CCL28 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CCL28
MM0153-9G34
Western Blot
chemokine (C-C motif) ligand 28, member 28, MGC71902, Protein CCK1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CCL28
Human CCL28
0.1 mL
Primary
56477