Learn More
CCL28 Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0179-7N12, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212116AF532
Catalog No. NP212116A32
Add to cart
Description
Description
CCL28 Monoclonal antibody specifically detects CCL28 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
CCL28
RM0179-7N12
Western Blot
chemokine (C-C motif) ligand 28, member 28, MGC71902, Protein CCK1
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CCL28
Mouse CCL28
0.1 mL
Primary
56477