Learn More
CCL3/MIP-1 alpha Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0154-11F16, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212118AF750
Catalog No. NP212118A70
Add to cart
Description
Description
CCL3/MIP-1 alpha Monoclonal antibody specifically detects CCL3/MIP-1 alpha in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
CCL3/MIP-1 alpha
MM0154-11F16
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78alpha, Mip1a, MIP-1alpha, MIP1-alpha, Scya3
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CCL3
Human CCL3
0.1 mL
Primary
6348